Tetrallyltin ≥96%

Supplier: Thermo Scientific Chemicals
Danger

71158-04 71158-09
CAAA71158-04EA 118.18 CAD
CAAA71158-04 CAAA71158-09
Tetrallyltin ≥96%
Tetrallyltin

MDL: MFCD00008637


Purity: 96% CAS No.: 7393-43-3 Molecular Formula: Sn(CH2CH=CH2)4 Formula Weight: 282.98 Flash Point: 75°(167°F) Physical Form: Liquid Density: 1.179 Refractive Index: 1.5385 Sensitivity: Air Sensitive Boiling Point: 69-70°/1.5mm UN No.: 2788 MDL No.: MFCD00008637

Formula: C₁₂H₂₀Sn
Boiling Pt: 69…70 °C (1,5 mmHg)
Density: 1,179
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008637
CAS Number: 7393-43-3
EINECS: 230-987-5
UN: 2788
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR