Phenyltin trichloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
71157-06
71157-14
CAAA71157-06EA
113.44
CAD
CAAA71157-06
CAAA71157-14
Phenyltin trichloride
Phenyltin trichloride
MDL: MFCD00000513
CAS No.: 1124-19-2
Molecular Formula: C6H5SnCl3
Formula Weight: 302.18
Physical Form: Liquid
Density: 1.84
Sensitivity: Moisture Sensitive
UN No.: 2788
MDL No.: MFCD00000513
Formula:
C₆H₅Cl₃Sn Boiling Pt: 109…110 °C (10 mmHg) Density: 1,84 Storage Temperature: Ambient |
MDL Number:
MFCD00000513 CAS Number: 1124-19-2 EINECS: 214-393-3 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...