Methyltin trichloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichloromethylstannane

71156-03 71156-06 71156-14
CAAA71156-03EA 115.97 CAD
CAAA71156-03 CAAA71156-06 CAAA71156-14
Methyltin trichloride 97%
Methyltin trichloride

MDL: MFCD00000514


Purity: 97% CAS No.: 993-16-8 Molecular Formula: CH3SnCl3 Formula Weight: 240.11 Physical Form: Crystalline Sensitivity: Moisture Sensitive UN No.: 2923 MDL No.: MFCD00000514

Formula: CH₃Cl₃Sn
MW: 240.1
Melting Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000514
CAS Number: 993-16-8
EINECS: 213-608-8
UN: 2921
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR