Tetraethyltin 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
71144-04
71144-09
71144-18
CAAA71144-04EA
68.89
CAD
CAAA71144-04
CAAA71144-09
CAAA71144-18
Tetraethyltin 98%
Tetraethyltin
MDL: MFCD00009020
Insoluble in water. Soluble in alcohol and ether
Purity: 98%
CAS No.: 597-64-8
Molecular Formula: (C2H5)4Sn
Formula Weight: 234.94
Flash Point: 53°(128°F)
Physical Form: Liquid
Density: 1.187
Refractive Index: 1.4730
Odor: Characteristic
Melting Point: -112°
Boiling Point: 181°
Solubility: Insoluble in water. Soluble in alcohol and ether
UN No.: 3384
MDL No.: MFCD00009020
Formula:
C₈H₂₀Sn Boiling Pt: 181 °C Melting Pt: –112 °C Density: 1,187 Flash Pt: 53 °C (128 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009020 CAS Number: 597-64-8 EINECS: 209-906-2 UN: 3384 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...