Dimethyltin dichloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
71142-04
71142-18
CAAA71142-04EA
54.19
CAD
CAAA71142-04
CAAA71142-18
Dimethyltin dichloride
Dimethyltin dichloride
MDL: MFCD00000517
Beilstein Registry No.: 3660508
CAS No.: 753-73-1
Molecular Formula: (CH3)2SnCl2
Formula Weight: 219.69
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 103-105°
Boiling Point: 185-190°
Beilstein Reference: 3660508
UN No.: 3146
MDL No.: MFCD00000517
Formula:
C₂H₆Cl₂Sn Boiling Pt: 185…190 °C Melting Pt: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000517 CAS Number: 753-73-1 EINECS: 212-039-2 UN: 3146 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...