Butyltin trichloride ≥96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
71125-14
71125-22
CAAA71125-14EA
52.14
CAD
CAAA71125-14
CAAA71125-22
Butyltin trichloride ≥96%
Butyltin trichloride
MDL: MFCD00000515
Purity: 96%
CAS No.: 1118-46-3
Molecular Formula: CH3(CH2)3SnCl3
Formula Weight: 282.17
Flash Point: 81°(178°F)
Physical Form: Liquid
Density: 1.693
Refractive Index: 1.5244
Sensitivity: Moisture Sensitive
Odor: Acrid
Melting Point: -63°
Boiling Point: 98°/10mm
UN No.: 3265
MDL No.: MFCD00000515
Formula:
C₄H₉Cl₃Sn Boiling Pt: 98 °C (10 mmHg) Melting Pt: –63 °C Density: 1,693 Flash Pt: 81 °C (178 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000515 CAS Number: 1118-46-3 EINECS: 214-263-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...