Tin(IV) bromide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
71120-18
71120-30
CAAA71120-18EA
180.12
CAD
CAAA71120-18
CAAA71120-30
Tin(IV) bromide 99%
Tin (IV) bromide
MDL: MFCD00011240
Purity: 99%
CAS No.: 7789-67-5
Molecular Formula: SnBr4
Formula Weight: 438.33
Physical Form: Lump
Density: 3.34
Appearance: Colorless to white
Odor: Acidic
Melting Point: 31°
Boiling Point: 202°/734mm
Merck Reference: 14,8773
UN No.: 3260
MDL No.: MFCD00011240
Formula:
Br₄Sn Boiling Pt: 202 °C (734 mmHg) Melting Pt: 31 °C Density: 3.34 Storage Temperature: Ambient |
MDL Number:
MFCD00011240 CAS Number: 7789-67-5 EINECS: 232-184-5 UN: 3260 ADR: 8,II Merck Index: 14,8773 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...