Tin(IV) iodide ≥95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tin tetraiodide

71114-09 71114-18
CAAA71114-09EA 103.81 CAD
CAAA71114-09 CAAA71114-18
Tin(IV) iodide ≥95%
Tin (IV) iodide

MDL: MFCD00036272 Soluble in alcohol, benzene, CS2, chloroform, and ether. Decomposes in water


Purity: 95% CAS No.: 7790-47-8 Molecular Formula: I4Sn Formula Weight: 626.31 Physical Form: Crystalline Density: 4.46 Refractive Index: 2.106 Sensitivity: Moisture Sensitive Appearance: Red Melting Point: ca 143° Boiling Point: 340° Solubility: Soluble in alcohol, benzene, CS2, chloroform, and ether. Decomposes in water Merck Reference: 14,8777 UN No.: 3260 MDL No.: MFCD00036272

Formula: I₄Sn
MW: 626.33 g/mol
Boiling Pt: 340 °C
Melting Pt: ∼ 143 °C
Density: 4.46
Storage Temperature: Ambient
MDL Number: MFCD00036272
CAS Number: 7790-47-8
EINECS: 232-208-4
UN: 3260
ADR: 8,III
Merck Index: 14,8777

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR