Tetrachlorosilane ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Silicon tetrachloride, Silicon (IV) chloride
69182-AB
69182-AE
69182-K2
69182-L7
CAAA69182-ABEA
36.81
CAD
CAAA69182-AB
CAAA69182-AE
CAAA69182-K2
CAAA69182-L7
Tetrachlorosilane ≥99% (metals basis)
Tetrachlorosilane
MDL: MFCD00011229
Notes: Decomposes in water with heat into silicic acid and HCl
Purity: 99%
CAS No.: 10026-04-7
Molecular Formula: Cl4Si
Formula Weight: 169.90
Physical Form: Liquid
Density: 1.483
Appearance: Colorless
Odor: Acrid
Melting Point: -70°
Boiling Point: 57.6°
Merck Reference: 14,8500
UN No.: 1818
MDL No.: MFCD00011229
Formula:
Cl₄Si MW: 169.9 g/mol Boiling Pt: 57.6 Melting Pt: -70 Density: 1.483 Storage Temperature: Ambient |
MDL Number:
MFCD00011229 CAS Number: 10026-04-7 EINECS: 233-054-0 UN: 1818 ADR: 8 II Merck Index: 14,8500 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...