Sodium hexafluorosilicate 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
69106-36
69106-A3
CAAA69106-36EA
89.27
CAD
CAAA69106-36
CAAA69106-A3
Sodium hexafluorosilicate 99+%
Sodium hexafluorosilicate
MDL: MFCD00003491
Slightly soluble in water. Solubility increases with temperature. Insoluble in alcohol
Purity: 99+%
CAS No.: 16893-85-9
Molecular Formula: F6Na2Si
Formula Weight: 188.06
Physical Form: Crystalline
Density: 2.679
Refractive Index: 1.310
Appearance: White
Melting Point: dec.
Solubility: Slightly soluble in water. Solubility increases with temperature. Insoluble in alcohol
Merck Reference: 14,8624
UN No.: 2674
MDL No.: MFCD00003491
Formula:
F₆Na₂Si Melting Pt: Dec. Density: 2.679 Storage Temperature: Ambient |
MDL Number:
MFCD00003491 CAS Number: 16893-85-9 EINECS: 240-934-8 UN: 2674 ADR: 6.1,III Merck Index: 14,8624 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...