Dimethyl selenide
Supplier: Thermo Scientific Chemicals
|
Danger
|
68116-03
68116-06
68116-14
CAAA68116-03EA
109.49
CAD
CAAA68116-03
CAAA68116-06
CAAA68116-14
Dimethyl selenide
Dimethyl selenide
MDL: MFCD00014848
Beilstein Registry No.: 1696848
CAS No.: 593-79-3
Molecular Formula: C2H6Se
Formula Weight: 109.03
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.41
Appearance: Colorless
Odor: Unpleasant
Boiling Point: 56-58°
Beilstein Reference: 1696848
UN No.: 3282
MDL No.: MFCD00014848
Formula:
C₂H₆Se MW: 109.03 g/mol Boiling Pt: 57 Density: 1.41 Storage Temperature: Refrigerator |
MDL Number:
MFCD00014848 CAS Number: 593-79-3 EINECS: 209-807-4 UN: 3282 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...