Potassium selenate ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

68111-04 68111-09 68111-18
CAAA68111-04EA 52.77 CAD
CAAA68111-04 CAAA68111-09 CAAA68111-18
Potassium selenate ≥99.5% (metals basis)
Potassium selenate

MDL: MFCD00054125 Soluble in water


Purity: 99.5% CAS No.: 7790-59-2 Molecular Formula: K2SeO4 Formula Weight: 221.16 Physical Form: -100 Mesh Powder Density: 3.07 Solubility: Soluble in water Merck Reference: 14,7666 UN No.: 2630 MDL No.: MFCD00054125

Formula: K₂O₄Se
Density: 3.07
Storage Temperature: Ambient
MDL Number: MFCD00054125
CAS Number: 7790-59-2
EINECS: 232-214-7
UN: 2630
ADR: 6.1,I
Merck Index: 14,7666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR