Selenium disulfide 97%

Supplier: Thermo Scientific Chemicals
Danger

68104-06 68104-18 68104-30
CAAA68104-06EA 60.99 CAD
CAAA68104-06 CAAA68104-18 CAAA68104-30
Selenium disulfide 97%
Selenium disulphide

MDL: MFCD00011227


Purity: 97% CAS No.: 7488-56-4 Molecular Formula: SeS2 Formula Weight: 143.09 Physical Form: Powder Melting Point: 100° Boiling Point: dec. Merck Reference: 14,8436 UN No.: 2657 MDL No.: MFCD00011227

Formula: S₂Se
Boiling Pt: Dec.
Melting Pt: <100 °C
Density: 0.03 g/cm³ (–65 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011227
CAS Number: 7488-56-4
EINECS: 231-303-8
UN: 2657
ADR: 6.1,II
Merck Index: 14,8436

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR