Scandium(III) nitrate hydate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
67104-03
67104-06
CAAA67104-03EA
202.24
CAD
CAAA67104-03
CAAA67104-06
Scandium(III) nitrate hydate ≥99.9% (REO, rare earth oxide basis), REacton®
Scandium (III) nitrate
MDL: MFCD00011220
Freely soluble in water and alcohol.
Purity: 99.9%
CAS No.: 13465-60-6
Molecular Formula: Sc(NO3)3.xH2O
Formula Weight: 230.97(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Solubility: Freely soluble in water and alcohol
Merck Reference: 14,8392
UN No.: 1477
MDL No.: MFCD00011220
Formula:
Sc(NO₃)₃·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00011220 CAS Number: 13465-60-6 EINECS: 236-701-5 UN: 1477 ADR: 5.1,III Merck Index: 14,8392 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...