Disodium sulfide, anhydrous
Supplier: Thermo Scientific Chemicals
|
Danger
|
In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagent
CAS No.: 1313-82-2
Molecular Formula: Na2S
Formula Weight: 78.04
Physical Form: Granular
Density: 1.856
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Pale yellow
Odor: Like rotten eggs
Melting Point: 950°
Solubility: Soluble in water. Decomposes in acid. Slightly soluble in alcohol. Insoluble in ether
Merck Reference: 14,8681
UN No.: 1385
MDL No.: MFCD00003498
In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagent
Formula:
Na₂S Boiling Pt: 174 °C (1013 hPa) Melting Pt: 950 °C Density: 1.856 Storage Temperature: Ambient |
MDL Number:
MFCD00003498 CAS Number: 1313-82-2 EINECS: 215-211-5 UN: 1385 ADR: 4.2,II REACH: 01-2119513694-38 Merck Index: 14,8681 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...