Rhenium trioxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

62109-14 62109-03 62109-06
CAAAA62109-14EA 3411.22 CAD
CAAAA62109-14 CAAA62109-03 CAAA62109-06
Rhenium trioxide ≥99.9% (metals basis)
Rhenium trioxide

MDL: MFCD00016306


Purity: 99.9% CAS No.: 1314-28-9 Molecular Formula: ReO3 Formula Weight: 236.20 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: Violet Boiling Point: 400° dec. Merck Reference: 14,8180 MDL No.: MFCD00016306

Formula: O₃Re
Boiling Pt: 400 °C (dec.)
Density: 7.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016306
CAS Number: 1314-28-9
EINECS: 215-228-8
Merck Index: 14,8180

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR