Sodium perrhenate ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
62107-03
62107-06
CAAA62107-03EA
110.44
CAD
CAAA62107-03
CAAA62107-06
Sodium perrhenate ≥99.95% (metals basis)
Sodium perrhenate
MDL: MFCD00014231
Purity: 99.95%
CAS No.: 13472-33-8
Molecular Formula: NaO4Re
Formula Weight: 273.19
Physical Form: Crystalline
Density: 5.390
Appearance: White
Odor: Odorless
Melting Point: 300°
UN No.: 1479
MDL No.: MFCD00014231
Formula:
NaO₄Re Melting Pt: 300 °C Density: 5,390 Storage Temperature: Ambient |
MDL Number:
MFCD00014231 CAS Number: 13472-33-8 EINECS: 236-742-9 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...