Lead(II) iodide ≥98.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lead diiodide
MDL: MFCD00011163
Partially soluble in water. Freely soluble in sodium thiosulfate solution. Soluble in concentrated solutions of alkali iodides. Insoluble in alcohol and cold HCl
Purity: 98.5%
CAS No.: 10101-63-0
Molecular Formula: I2Pb
Formula Weight: 461.00
Physical Form: Powder
Density: 6.16
Appearance: Yellow
Odor: Odorless
Melting Point: 402°
Boiling Point: 954°
Solubility: Partially soluble in water. Freely soluble in sodium thiosulfate solution. Soluble in concentrated solutions of alkali iodides. Insoluble in alcohol and cold HCl
Merck Reference: 14,5411
UN No.: 2291
MDL No.: MFCD00011163
Formula:
I₂Pb MW: 461.01 g/mol Boiling Pt: 954 Melting Pt: 402 Density: 6.16 Storage Temperature: Ambient |
MDL Number:
MFCD00011163 CAS Number: 10101-63-0 EINECS: 233-256-9 UN: 2291 ADR: 6.1 III Merck Index: 14,5411 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...