Dodecamolybdophosphoric acid hydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

56166-06 56166-14 56166-22 56166-36
CAAA56166-06EA 50.4 CAD
CAAA56166-06 CAAA56166-14 CAAA56166-22 CAAA56166-36
Dodecamolybdophosphoric acid hydrate ACS
Dodecamolybdophosphoric acid hydrate

MDL: MFCD00149913


CAS No.: 51429-74-4 Molecular Formula: H3PO4.12MoO3.xH2O Formula Weight: 1825.25(anhy) Physical Form: Granular Density: 3.15 Sensitivity: Moisture Sensitive Appearance: Yellow Melting Point: 78-90° Merck Reference: 14,7342 UN No.: 3085 MDL No.: MFCD00149913

Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
Melting Pt: 78…90 °C
Density: 3.15
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,7342

Order Now

Specification Test Results

Insoluble matter 0.01
Cl 0.02
SO3 0.025
NH3 0.01%
Ca 0.02%
Heavy Metals (as Pb) 0.005%
Fe 0.00005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR