Sodium thiophosphate hydrate

Supplier: Thermo Scientific Chemicals
Warning

56146-06 56146-14
CAAA56146-06EA 156.1 CAD
CAAA56146-06 CAAA56146-14
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate

MDL: MFCD00041513


CAS No.: 10489-48-2 Molecular Formula: Na3PO3S.xH2O Formula Weight: 179.93(anhy) Physical Form: Crystalline aggregates Appearance: White to green Melting Point: 53-58° MDL No.: MFCD00041513

Formula: Na₂S₂O₃
Melting Pt: 53…58 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041513
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 14,8693

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR