Phosphoryl tribromide ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
56135-06
56135-14
56135-22
CAAA56135-06EA
73.94
CAD
CAAA56135-06
CAAA56135-14
CAAA56135-22
Phosphoryl tribromide ≥98%
Phosphoryl tribromide
As a chemical intermediate
Purity: 98% min
CAS No.: 7789-59-5
Molecular Formula: POBr3
Formula Weight: 286.73
Physical Form: Lump
Density: 2.822
Melting Point: 56°
Boiling Point: 189°
Solubility: Decomposes in water. Soluble in ether, benzene, and sulfuric acid
Merck Reference: 14,7348
UN No.: 1939
MDL No.: MFCD00036298
As a chemical intermediate
Formula:
Br₃OP Boiling Pt: 189 °C Melting Pt: 56 °C Density: 2.822 Storage Temperature: Ambient |
MDL Number:
MFCD00036298 CAS Number: 7789-59-5 EINECS: 232-177-7 UN: 1939 ADR: 8,II Merck Index: 14,7348 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...