Ammonium persulfate (APS) ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Oxidizer, bleaching agent, photography, aniline dyes, electroplating, in analytical chemistry for determination of manganese
Purity: 98.0% min
CAS No.: 7727-54-0
Molecular Formula: H8N2O8S2
Formula Weight: 228.19
Flash Point: None
Physical Form: Lump
Density: 1.982
Refractive Index: 1.50
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Odorless
Melting Point: 120° dec.
Solubility: Freely soluble in water
Merck Reference: 14,541
UN No.: 1444
MDL No.: MFCD00003390
Oxidizer, bleaching agent, photography, aniline dyes, electroplating, in analytical chemistry for determination of manganese
Formula:
(NH₄)₂S₂O₈ Melting Pt: 120 °C (dec.) Density: 1.982 Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 14,541 |
|
Specification Test Results
Insoluble matter | 0.005% |
Residue after ignition | 0.05% |
Titratable free acid | 0.04meq/g |
Chloride and chlorate (as Cl) | 0.00001 |
Heavy Metals (as Pb) | 0.005% |
Fe | 0.001% |
Mn | 0.5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...