Nickel(II) chloride hexahydrate ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickel dichloride hexahydrate
53131-18
53131-30
53131-A1
CAAA53131-18EA
84.37
CAD
CAAA53131-18
CAAA53131-30
CAAA53131-A1
Nickel(II) chloride hexahydrate ≥99.95% (metals basis)
Nickel (II) chloride hexahydrate
MDL: MFCD00149809
Fieser: 12,336 15,217 21,281
Purity: 99.95%
CAS No.: 7791-20-0
Molecular Formula: Cl2Ni.6H2O
Formula Weight: 237.71 (129.60anhy)
Physical Form: Crystalline
Density: 1.92
Appearance: Green
Odor: Odorless
pH: 4.9
Melting Point: 140° -H2O
Merck Reference: 14,6505
UN No.: 3288
MDL No.: MFCD00149809
Formula:
NiCl₂·6H₂O Melting Pt: 140 °C (-H₂O) Density: 1.92 Storage Temperature: Ambient |
MDL Number:
MFCD00149809 CAS Number: 7791-20-0 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III Merck Index: 14,6505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...