Nickel (II) sulfate hexahydrate ≥99.97% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
53130-18
53130-30
CAAA53130-18EA
135.56
CAD
CAAA53130-18
CAAA53130-30
Nickel (II) sulfate hexahydrate ≥99.97% (metals basis)
Nickel sulphate hexahydrate
MDL: MFCD00149813
Notes: m.p. 100°C -5H2O, 280°C -6H2O
Purity: 99.97% min
CAS No.: 10101-97-0
Molecular Formula: NiO4S.6H2O
Formula Weight: 262.86 (154.77anhy)
Physical Form: Lump
Density: 2.070
Appearance: Green
Odor: Odorless
pH: 4.3-4.7
Merck Reference: 14,6517
UN No.: 3288
MDL No.: MFCD00149813
Formula:
NiSO₄·6H₂O Melting Pt: 840 °C Density: 2.070 Storage Temperature: Ambient |
MDL Number:
MFCD00149813 CAS Number: 10101-97-0 EINECS: 232-104-9 UN: 3288 ADR: 6.1,III Merck Index: 14,6517 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...