Nickel(II) bromide, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickel dibromide
53115-18
53115-30
CAAA53115-18EA
170.64
CAD
CAAA53115-18
CAAA53115-30
Nickel(II) bromide, anhydrous 99%
Nickel (II) bromide
MDL: MFCD00011141
Purity: 99%
CAS No.: 13462-88-9
Molecular Formula: Br2Ni
Formula Weight: 218.53
Physical Form: Crystalline
Density: 5.098
Melting Point: 963°
Merck Reference: 14,6502
UN No.: 3077
MDL No.: MFCD00011141
Formula:
Br₂Ni MW: 218.5 g/mol Melting Pt: 963 Density: 5.098 Storage Temperature: Ambient |
MDL Number:
MFCD00011141 CAS Number: 13462-88-9 EINECS: 236-665-0 UN: 3077 ADR: 9 III Merck Index: 14,6502 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...