Molybdenum dioxide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals

48117-14 48117-22
CAAA48117-14EA 84.06 CAD
CAAA48117-14 CAAA48117-22
Molybdenum dioxide ≥99% (metals basis)
Molybdenum dioxide

MDL: MFCD00014223


Purity: 99% CAS No.: 18868-43-4 Molecular Formula: MoO2 Formula Weight: 127.94 Physical Form: Powder Density: 6.47 Appearance: Brown to black Odor: Odorless MDL No.: MFCD00014223

Formula: MoO₂
Boiling Pt: 1207…1307 °C (1013 hPa)
Melting Pt: 770…795 °C
Density: 6,47
Storage Temperature: Ambient
MDL Number: MFCD00014223
CAS Number: 18868-43-4
EINECS: 242-637-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR