Potassium molybdate(VI) ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassium molybdate, Molybdic acid potassium salt, Dipotassium tetraoxomolybdate, Molybdic acid, potassium salt, Potassium molybdenum oxide, di-Potassium molybdate
MDL: MFCD00011370
Soluble in water. Insoluble in alcohol
Purity: 95%
CAS No.: 13446-49-6
Molecular Formula: K2MoO4
Formula Weight: 238.14
Physical Form: Powder
Density: 2.3
Melting Point: 919°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,7647
MDL No.: MFCD00011370
Formula:
K₂MoO₄ Melting Pt: 919 °C Density: 2.3 Storage Temperature: Ambient |
MDL Number:
MFCD00011370 CAS Number: 13446-49-6 EINECS: 236-599-2 Merck Index: 14,7647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...