Magnesium bromide hexahydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
46120-22
46120-36
46120-A3
CAAA46120-22EA
80.42
CAD
CAAA46120-22
CAAA46120-36
CAAA46120-A3
Magnesium bromide hexahydrate 98+%
Magnesium bromide hexahydrate
MDL: MFCD00149780
Purity: 98+%
CAS No.: 13446-53-2
Molecular Formula: Br2Mg.6H2O
Formula Weight: 292.22 (184.13anhy)
Physical Form: Crystalline
Density: 2.00
Appearance: Colorless
Odor: Odorless
Melting Point: 172°
Solubility: Soluble in water, somewhat soluble in alcohol
MDL No.: MFCD00149780
Formula:
Br₂Mg·6H₂O Melting Pt: 172 °C Density: 2,00 Storage Temperature: Ambient |
MDL Number:
MFCD00149780 CAS Number: 13446-53-2 EINECS: 232-170-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...