Magnesium thiosulfate hexahydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
46114-22
46114-36
CAAA46114-22EA
75.05
CAD
CAAA46114-22
CAAA46114-36
Magnesium thiosulfate hexahydrate 99%
Magnesium thiosulphate hexahydrate
MDL: MFCD00169871
Soluble in water. Insoluble in alcohol
Purity: 99%
CAS No.: 13446-30-5
Molecular Formula: MgO3S2.6H2O
Formula Weight: 244.53 (136.44anhy)
Physical Form: Crystalline
Density: 1.818
Appearance: White
Melting Point: 170° (-3H2O)
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,5694
MDL No.: MFCD00169871
Formula:
MgO₃S₂·6H₂O Melting Pt: 170 °C (-3H₂O) Density: 1.818 Storage Temperature: Ambient |
MDL Number:
MFCD00169871 CAS Number: 13446-30-5 EINECS: 233-340-5 Merck Index: 14,5694 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...