3-Hydroxy-5-methylisoxazole 97%

Supplier: Thermo Scientific Chemicals
Danger

44450-06 44450-14 44450-22
CA44450-06EA 84.69 CAD
CA44450-06 CAAA44450-14 CAAA44450-22
3-Hydroxy-5-methylisoxazole 97%
3-Hydroxy-5-methylisoxazole

MDL: MFCD00144468 Soluble in alcohol, acetone, THF, chloroform


Purity: 97% CAS No.: 10004-44-1 Molecular Formula: C4H5NO2 Formula Weight: 99.09 Physical Form: Powder Solubility: Soluble in alcohol, acetone, THF, chloroform MDL No.: MFCD00144468

Formula: C₄H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00144468
CAS Number: 10004-44-1
EINECS: 233-000-6
Merck Index: 14,4856

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR