Manganese(II) bromide tetrahydrate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Manganous bromide tetrahydrate, Manganese dibromide tetrahydrate, Manganese(II) bromide hydrate
44443-09
44443-18
CAAA44443-09EA
127.51
CAD
CAAA44443-09
CAAA44443-18
Manganese(II) bromide tetrahydrate ≥99.9% (metals basis)
Manganese (II) bromide tetrahydrate
MDL: MFCD00149791
Purity: 99.9%
CAS No.: 10031-20-6
Molecular Formula: MnBr2.xH2O
Formula Weight: 214.76(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Pink
Odor: Odorless
Melting Point: 64.3° dec.
Merck Reference: 14,5725
MDL No.: MFCD00149791
Formula:
Br₂Mn·4H₂O MW: 286.81 g/mol Melting Pt: 64.3 °C (dec.) Density: 4.38 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149791 CAS Number: 10031-20-6 EINECS: 236-591-9 Merck Index: 14,5725 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...