Manganese(II) chloride tetrahydrate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

44442-06 44442-14 44442-22
CAAA44442-06EA 66.52 CAD
CAAA44442-06 CAAA44442-14 CAAA44442-22
Manganese(II) chloride tetrahydrate ≥99.99% (metals basis)
Manganese (II) chloride tetrahydrate

MDL: MFCD00149792


Purity: 99.99% CAS No.: 13446-34-9 Molecular Formula: MnCl2.4H2O Formula Weight: 197.90 (125.84anhy) Physical Form: Crystalline Density: 2.010 Melting Point: 58° Merck Reference: 14,5728 MDL No.: MFCD00149792

Formula: MnCl₂·4H₂O
MW: 197.9 g/mol
Boiling Pt: 1190 °C (1013 hPa)
Melting Pt: 58 °C
Density: 2.010
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 14,5728

Order Now

Specification Test Results

Form Crystalline

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR