Manganese(II) oxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44441-09
44441-18
CAAA44441-09EA
249.64
CAD
CAAA44441-09
CAAA44441-18
Manganese(II) oxide ≥99.99% (metals basis)
Manganese (II) oxide
MDL: MFCD00016226
Purity: 99.99%
CAS No.: 1344-43-0
Molecular Formula: MnO
Formula Weight: 70.94
Physical Form: Powder/Chunks
Density: 5.0
Appearance: Green
Odor: Odorless
MDL No.: MFCD00016226
Formula:
MnO Melting Pt: 1840 °C Density: 5.0 Storage Temperature: Ambient |
MDL Number:
MFCD00016226 CAS Number: 1344-43-0 EINECS: 215-695-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...