Phosphorus(III) chloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phosphorus trichloride
44436-18
44436-30
CAAA44436-18EA
56.88
CAD
CAAA44436-18
CAAA44436-30
Phosphorus(III) chloride ≥98%
Phosphorus (III) chloride
MDL: MFCD00011438
Purity: 98% min
CAS No.: 7719-12-2
Molecular Formula: PCl3
Formula Weight: 137.33
Physical Form: Liquid
Density: 1.574
Refractive Index: 1.516
Odor: Acrid
Melting Point: -111.8°
Boiling Point: 76°
Merck Reference: 14,7358
UN No.: 1809
MDL No.: MFCD00011438
Formula:
Cl₃P MW: 137.33 g/mol Boiling Pt: 76 Melting Pt: -112 Density: 1.574 Storage Temperature: Ambient |
MDL Number:
MFCD00011438 CAS Number: 7719-12-2 EINECS: 231-749-3 UN: 1809 ADR: 6.1 I Merck Index: 14,7358 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...