Titanium(IV) oxide anatase catalyst support

Supplier: Thermo Scientific Chemicals
Warning

44429-30 44429-A1 44429-A7
CAAA44429-30EA 122.45 CAD
CAAA44429-30 CAAA44429-A1 CAAA44429-A7
Titanium(IV) oxide anatase catalyst support
Titanium (IV) oxide anatase

MDL: MFCD00011269 Notes: Anatase, Typical properties= S.A. greater than 150m2/g, median pore diameter 140┼, total pore volume 0.38cc/g, packing density 0.90kg/L, contains 1% S (as SO4)


Molecular Formula: TiO2 Formula Weight: 79.90 Physical Form: 1/8" Pellets MDL No.: MFCD00011269

Formula: TiO₂
Boiling Pt: 2900 °C (1013 hPa)
Melting Pt: 1855 °C
Density: 4.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 215-280-1
Merck Index: 13,09549

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR