Lutetium trifluoride ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

44425-03 44425-06
CAAA44425-03EA 164.32 CAD
CAAA44425-03 CAAA44425-06
Lutetium trifluoride ≥99.9% (metals basis)
Lutetium trifluoride

MDL: MFCD00016191


Purity: 99.9% CAS No.: 13760-81-1 Molecular Formula: LuF3 Formula Weight: 231.97 Physical Form: -325 Mesh Powder Density: 8.3 Melting Point: 1182° UN No.: 3288 MDL No.: MFCD00016191

Formula: LuF₃
Boiling Pt: 2200 °C (1013 hPa)
Melting Pt: 1182 °C
Density: 8,3
Storage Temperature: Ambient
MDL Number: MFCD00016191
CAS Number: 13760-81-1
EINECS: 237-355-8
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR