Potassium dihydrogen citrate hydrate ≥99% (dry basis)

Supplier: Thermo Scientific Chemicals

44415-36 44415-A3
CAAA44415-36EA 143.46 CAD
CAAA44415-36 CAAA44415-A3
Potassium dihydrogen citrate hydrate ≥99% (dry basis)
Potassium dihydrogen citrate hydrate

Potassium dihydrogen citrate hydrate is used as a buffering agent and a chelating agent. It is used as an alkalizing agent in the renal function. It is found that it increases the bone density, thereby used in research experiments related to the treatment of osteoporosis.


Purity: 99% CAS No.: 866-83-1 Molecular Formula: HOOCCH2C(OH)(COOH)CH2COOK.xH2O Formula Weight: 230.22(anhy) Physical Form: Powder/ chunks Sensitivity: Hygroscopic Beilstein Reference: 4772973 Merck Reference: 14,7624 MDL No.: MFCD00036472

Formula: C₆H₇KO₇
Storage Temperature: Ambient
MDL Number: MFCD00036472
CAS Number: 866-83-1
EINECS: 212-753-4
Merck Index: 14,7624

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR