Lanthanum(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

44396-09 44396-18
CAAA44396-09EA 71.42 CAD
CAAA44396-09 CAAA44396-18
Lanthanum(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Lanthanum (III) bromide

MDL: MFCD00016161


Purity: 99.9% CAS No.: 13536-79-3 Molecular Formula: LaBr3 Formula Weight: 378.62 Physical Form: Lumps, Ampouled under argon Sensitivity: Hygroscopic Appearance: White MDL No.: MFCD00016161

Formula: Br₃La
MW: 378.62
Melting Pt: 789
Density: 5.06
Storage Temperature: Ambient
MDL Number: MFCD00016161
CAS Number: 13536-79-3
EINECS: 236-896-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR