Silica gel, granules -6+12 mesh desiccant

Supplier: Thermo Scientific Chemicals

44387-30 44387-A1 44387-A7
CAAA44387-30EA 44.24 CAD
CAAA44387-30 CAAA44387-A1 CAAA44387-A7
Silica gel, granules -6+12 mesh desiccant
Silica gel

MDL: MFCD00011232 Notes: Re-use after being saturated with water by heating to 121 °C for approximately 8 hours


CAS No.: 7631-86-9 Sensitivity: Hygroscopic Odor: Odorless MDL No.: MFCD00011232

Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
REACH: 01-2119379499-16

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR