Silica gel, granules -3/+8 mesh desiccant

Supplier: Thermo Scientific Chemicals

44386-30 44386-A1 44386-A7
CAAA44386-30EA 45.35 CAD
CAAA44386-30 CAAA44386-A1 CAAA44386-A7
Silica gel, granules -3/+8 mesh desiccant
Silica gel

MDL: MFCD00011232 Notes: Re-use after being saturated with water by heating to 121 °C for approximately 8 hours


CAS No.: 7631-86-9 Sensitivity: Hygroscopic Odor: Odorless MDL No.: MFCD00011232

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
REACH: 01-2119379499-16

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR