Cerium(III) sulfate octahydrate ≥99.999% (metals basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dicerium trisulfate octahydrate, Dicerium trisulphate octahydrate

44357-18 44357-30
CAAA44357-18EA 263.86 CAD
CAAA44357-18 CAAA44357-30
Cerium(III) sulfate octahydrate ≥99.999% (metals basis), REacton®
Cerium (III) sulphate octahydrate

MDL: MFCD00149641


Purity: 99.999% CAS No.: 10450-59-6 Molecular Formula: Ce2(SO4)3.8H2O Formula Weight: 712.55 (568.42anhy) Physical Form: Crystalline Density: 2.886 Merck Reference: 14,2002 MDL No.: MFCD00149641

Formula: Ce₂O₁₂S₃·8H₂O
MW: 712.54 g/mol
Density: 2.886
Storage Temperature: Ambient
MDL Number: MFCD00149641
CAS Number: 10450-59-6
EINECS: 236-644-6
Merck Index: 14,2002

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR