Copper(II) acetate ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid copper (II) salt, Cupric acetate, Copper diacetate

44355-06 44355-14 44355-22
CAAA44355-06EA 169.06 CAD
CAAA44355-06 CAAA44355-14 CAAA44355-22
Copper(II) acetate ≥99.999% (metals basis)
Copper (II) acetate

MDL: MFCD00008690


Purity: 99.999% CAS No.: 142-71-2 Molecular Formula: Cu(OOCCH3)2 Formula Weight: 181.63 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Blue-green Odor: Odorless Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00008690

Formula: CuCH₃CO₂
MW: 181.64 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008690
CAS Number: 142-71-2
EINECS: 205-553-3
UN: 3077
ADR: 9,III
Merck Index: 14,2624

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR