Lithium sulfate, anhydrous ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

44352-14 44352-22
CAAA44352-14EA 216.46 CAD
CAAA44352-14 CAAA44352-22 CAAA44352-36
Lithium sulfate, anhydrous ≥99.99% (metals basis)
Lithium sulphate

MDL: MFCD00011086


Purity: 99.99% CAS No.: 10377-48-7 Molecular Formula: Li2SO4 Formula Weight: 109.94 Physical Form: Coarse Powder Density: 2.22 Appearance: White Odor: Odorless Melting Point: 845° Merck Reference: 14,5541 MDL No.: MFCD00011086

Formula: Li₂O₄S
MW: 109.95 g/mol
Boiling Pt: 1377 °C (1013 hPa)
Melting Pt: 845 °C
Density: 2.22
Storage Temperature: Ambient
MDL Number: MFCD00011086
CAS Number: 10377-48-7
EINECS: 233-820-4
Merck Index: 14,5541

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR