Iodine monochloride ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iodochloride

44348-14 44348-22 44348-36
CAAA44348-14EA 96.06 CAD
CAAA44348-14 CAAA44348-22 CAAA44348-36
Iodine monochloride ACS
Iodine monochloride

MDL: MFCD00011354 Beilstein Registry No.: 3902972 Fieser: 1,502 8,261 10,212 11,268 18,193 19,176 20,200 21,233


CAS No.: 7790-99-0 Molecular Formula: ClI Formula Weight: 162.36 Physical Form: Crystalline/Liquid Density: 3.182 Sensitivity: Moisture and Light Sensitive Appearance: Dark red/black Odor: Pungent Melting Point: 25-27° Boiling Point: 96-98° Beilstein Reference: 3902972 Merck Reference: 14,5017 UN No.: 1792 MDL No.: MFCD00011354

Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 97.3
Melting Pt: 26
Density: 3.182
Storage Temperature: Ambient
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 1792
ADR: 8 II
Merck Index: 14,5017

Order Now

Specification Test Results

Assay I/Cl ratio of the Wijs solution prepared from the reagent must be 1.10+/-0.1
Insoluble matter 0.005% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR