Lanthanum(III) nitrate hexahydrate ≥99.999% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

44346-14 44346-22 44346-36
CAAA44346-14EA 136.2 CAD
CAAA44346-14 CAAA44346-22 CAAA44346-36
Lanthanum(III) nitrate hexahydrate ≥99.999% (REO, rare earth oxide basis), REacton®
Lanthanum (III) nitrate hexahydrate

MDL: MFCD00149751


Purity: 99.999% CAS No.: 10277-43-7 Molecular Formula: La(NO3)3.6H2O Formula Weight: 433.01 (324.92anhy) Physical Form: Crystalline Odor: Odorless Merck Reference: 14,5363 UN No.: 1477 MDL No.: MFCD00149751

Formula: La(NO₃)₃·6H₂O
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: 40 °C
Density: 2.347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1,III
Merck Index: 14,5363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR