Aluminium 99,9999 Metals basis, foil, Puratronic®, annealed, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

44331-FI 44331-GH 44331-HP
CAAA44331-FIEA 122.92 CAD
CAAA44331-FI CAAA44331-GH CAAA44331-HP
Aluminium 99,9999 Metals basis, foil, Puratronic®, annealed, Thickness: 0.25 mm (0.01 in)
Aluminium

MDL: MFCD00134029


Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈1.7g/50x50mm, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR