Aluminium ≥99.9999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.5 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
44329-09
44329-18
CAAA44329-09EA
143.62
CAD
CAAA44329-09
CAAA44329-18
Aluminium ≥99.9999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.5 in), Ø 6.35 mm (0.25 in)
Aluminium
Purity: 99.9999%
CAS No.: 7429-90-5
Physical Form: ≈1.1g/slug
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2327 Melting Pt: 660 Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...