Aluminium ≥99.9999% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
44328-06
44328-14
44328-22
CAAA44328-06EA
108.23
CAD
CAAA44328-06
CAAA44328-14
CAAA44328-22
Aluminium ≥99.9999% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Aluminium
MDL: MFCD00134029
Purity: 99.9999%
CAS No.: 7429-90-5
Physical Form: ≈0.54g/slug
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...