Lanthanum(III) chloride, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44322-06
44322-14
CAAA44322-06EA
118.82
CAD
CAAA44322-06
CAAA44322-14
Lanthanum(III) chloride, ultra dry ≥99.99% (metals basis)
Lanthanum (III) chloride
MDL: MFCD00011068
Purity: 99.99%
CAS No.: 10099-58-8
Molecular Formula: LaCl3
Formula Weight: 245.27
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.842
Appearance: White
Odor: Odorless
Melting Point: 860°
Boiling Point: 1000°
Merck Reference: 14,5363
UN No.: 3077
MDL No.: MFCD00011068
Formula:
Cl₃La Boiling Pt: 1000 °C Melting Pt: 860 °C Density: 3.842 Storage Temperature: Ambient |
MDL Number:
MFCD00011068 CAS Number: 10099-58-8 EINECS: 233-237-5 UN: 3077 ADR: 9,III Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...