Cobalt(II) perchlorate hexahydrate ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cobalt diperchlorate hexahydrate
44320-09
44320-18
CAAA44320-09EA
184.86
CAD
CAAA44320-09
CAAA44320-18
CAAA44320-30
Cobalt(II) perchlorate hexahydrate ≥99.999% (metals basis)
Cobalt (II) perchlorate hexahydrate
MDL: MFCD00150207
Purity: 99.999%
CAS No.: 13478-33-6
Molecular Formula: Co(ClO4)2.6H2O
Formula Weight: 366.74 (257.83anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
UN No.: 1481
MDL No.: MFCD00150207
Formula:
Cl₂CoO₈·6H₂O MW: 365.93 Storage Temperature: Ambient |
MDL Number:
MFCD00150207 CAS Number: 13478-33-6 EINECS: 236-653-5 UN: 1481 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...